Learn More
Braintree Scientific Vertical Charcoal Canister Adapter for F/Air Canister only
SDP
Catalog No. 501954838
Encompass
Vertical Charcoal Canister Adapter connects standard 1/4 tubing to the F/Aircanister. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4838 Supplier Braintree Scientific Supplier No. FCACONNECT
Vertical Charcoal Canister Adapter connects standard 1/4 tubing to the F/Aircanister. **Sold for research purposes only and not for human use**