Learn More
Bellco Glass, Inc. Vertical Disk Impeller, 3L Bioreactor - 2 1/4 OD"
SDP
Catalog No. 501944728
Encompass_Preferred
VERTICAL DISK IMPELLER 3L MADE IN THE USA Material: 316 SSTFor bioreactor system - 7802-03115Dimensions: 2" od x 47/64" wideFor .375" od shafts
Catalog No. 50-194-4728 Supplier Bellco Glass, Inc. Supplier No. 196402080
VERTICAL DISK IMPELLER 3L MADE IN THE USA Material: 316 SSTFor bioreactor system - 7802-03115Dimensions: 2" od x 47/64" wideFor .375" od shafts