Learn More
Bellco Glass, Inc. Vertical Disk Impeller,7L For 7L Bioreactor Systems
SDP
Catalog No. 501944735
Encompass_Preferred
For 7l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finishOD: 2 3/4"Requires two (2) ea per vessel
Catalog No. 50-194-4735 Supplier Bellco Glass, Inc. Supplier No. 196402300
For 7l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finishOD: 2 3/4"Requires two (2) ea per vessel