Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Half (Slim) Rack for 2" boxes, Holds 3 boxes
SDP
Catalog No. 503028683
Encompass_Preferred
For Standard 2″ Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 2 inch boxes. These slim style vertical freezer racks allow you to maximize your freezers capacity storage by filling in gaps that can’t fit standard chest/LN2 freezer racks. Each slim style vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x D x W: 16 1/2 x 2 1/4 x 5 9/16 (419 x 57 x 141mm)Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 (137 x 54.4 x 133mm)  
Catalog No. 50-302-8683 Supplier Alkali Scientific Supplier No. LR32
For Standard 2″ Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 2 inch boxes. These slim style vertical freezer racks allow you to maximize your freezers capacity storage by filling in gaps that can’t fit standard chest/LN2 freezer racks. Each slim style vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x D x W: 16 1/2 x 2 1/4 x 5 9/16 (419 x 57 x 141mm)Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 (137 x 54.4 x 133mm)  