Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for 100-Place Slide Boxes, Holds 6 boxes
SDP
Catalog No. 503029381
Encompass_Preferred
For 100-Place Slide Boxes The CFS-100-6 vertical freezer racks for storage of 100-place slide boxes, features a 6 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and also features locking rods for added security. This vertical freezer rack does not come standard with 100-place slide boxes. Dimensions L x H x W : 9 1/2 x 7 5/16 x 8 11/16 inch (242 x 185 x 220 mm)Compartment Dimensions h x d x w: 1 9/16 x 7 1/8 x 8 3/8 inch (38.5 x 181 x 212mm) *Security lock device is ordered separately.
Catalog No. 50-302-9381 Supplier Alkali Scientific Supplier No. CFS1006
For 100-Place Slide Boxes The CFS-100-6 vertical freezer racks for storage of 100-place slide boxes, features a 6 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and also features locking rods for added security. This vertical freezer rack does not come standard with 100-place slide boxes. Dimensions L x H x W : 9 1/2 x 7 5/16 x 8 11/16 inch (242 x 185 x 220 mm)Compartment Dimensions h x d x w: 1 9/16 x 7 1/8 x 8 3/8 inch (38.5 x 181 x 212mm) *Security lock device is ordered separately.