Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for 56 CBBC-V1 Canisters
SDP
Catalog No. 503026515
Encompass_Preferred
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific  Canister and includes a handle. Stainless Steel Vertical Rack for 56CBBC-V1 Canisters Blood Bag Number:791-OS791-OSU8-0346-O Dimensions (D x H x L): 3 9/11 x 25 x 4 3/8″97 x 635 x 111 mm Canister: CBBC-V1 Platform DividersP20-CBBC-VP24-CBBC-VP31-CBBC-VP39-CBBC-V **Lock device is ordered separately
Catalog No. 50-302-6515 Supplier Alkali Scientific Supplier No. CF83T
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific  Canister and includes a handle. Stainless Steel Vertical Rack for 56CBBC-V1 Canisters Blood Bag Number:791-OS791-OSU8-0346-O Dimensions (D x H x L): 3 9/11 x 25 x 4 3/8″97 x 635 x 111 mm Canister: CBBC-V1 Platform DividersP20-CBBC-VP24-CBBC-VP31-CBBC-VP39-CBBC-V **Lock device is ordered separately