Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for 63 CBBC-V1 Canisters
SDP
Catalog No. 503026548
Encompass_Preferred
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Vertical Rack for 63CBBC-V1 Canisters Blood Bag Number:791-OS791-OSU8-0346-O Dimensions (D x H x L): 3 9/11 x 28 1/16 x 4 3/8″97 x 713 x 111mm Canister: CBBC-V1 Platform DividersP20-CBBC-VP24-CBBC-VP31-CBBC-VP39-CBBC-V
Catalog No. 50-302-6548 Supplier Alkali Scientific Supplier No. CF93T
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Vertical Rack for 63CBBC-V1 Canisters Blood Bag Number:791-OS791-OSU8-0346-O Dimensions (D x H x L): 3 9/11 x 28 1/16 x 4 3/8″97 x 713 x 111mm Canister: CBBC-V1 Platform DividersP20-CBBC-VP24-CBBC-VP31-CBBC-VP39-CBBC-V