Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for Mini 3" boxes, Holds 10 boxes
SDP
Catalog No. 503028472
Encompass_Preferred
For Standard 3″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 3 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x Wx D: 32 1/16 x 3 3/8 x 3 1/4 inch (814 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.  
Catalog No. 50-302-8472 Supplier Alkali Scientific Supplier No. MCF103
For Standard 3″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 3 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x Wx D: 32 1/16 x 3 3/8 x 3 1/4 inch (814 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.  