Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for Mini 3" boxes, Holds 6 boxes
SDP
Catalog No. 503029472
Encompass_Preferred
For Standard 3″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 3 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x Wx D: 19 3/8 x 3 3/8 x 3 1/4 inch (492 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.  
Catalog No. 50-302-9472 Supplier Alkali Scientific Supplier No. MCF63
For Standard 3″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 3 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x Wx D: 19 3/8 x 3 3/8 x 3 1/4 inch (492 x 86 x 82mm)Compartment Dimensions H x W x D: 3 1/8 x 3 1/16 x 3 1/16 inch (80 x 78 x 78mm) **Lock device is ordered separately.  