Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for Mini 2" boxes, Holds 13 boxes
SDP
Catalog No. 503026724
Encompass_Preferred
For Standard 2″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 2 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x W x D : 28 1/2 x 3 3/8 x 3 1/4 ( 724 x 86 x 82mm)Compartment Dimensions: 2 3/16 x 3 1/16 x 3 1/16 ( 55 x 78 x 78mm) MCF-13-2-X: Rack w/ 2″ Box and Divider *To order replace X with B-Cardboard Mini Box with 25 cell divider or P-Mini Polycarbonate Box with 25 cell divider **Lock device is ordered separately. ***w2= 0.09lb(0.04kg) for cardboard box, 0.37lb(0.17kg) for plastic box.    
Catalog No. 50-302-6724 Supplier Alkali Scientific Supplier No. MCF132
For Standard 2″ Mini Boxes Our vertical freezer half racks for laboratory chest and liquid nitrogen freezers provides storage of 2 inch mini boxes. These vertical freezer racks allow you to maximize your freezers capacity storage. Each vertical rack is built out of corrosion resistant stainless steel. Each freezer rack includes handles allowing for an easy retrieval from a chest or LN2 freezer and locking rods for added security.  Dimensions H x W x D : 28 1/2 x 3 3/8 x 3 1/4 ( 724 x 86 x 82mm)Compartment Dimensions: 2 3/16 x 3 1/16 x 3 1/16 ( 55 x 78 x 78mm) MCF-13-2-X: Rack w/ 2″ Box and Divider *To order replace X with B-Cardboard Mini Box with 25 cell divider or P-Mini Polycarbonate Box with 25 cell divider **Lock device is ordered separately. ***w2= 0.09lb(0.04kg) for cardboard box, 0.37lb(0.17kg) for plastic box.    