Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for SBS Formatted Boxes, Holds 6 Boxes
SDP
Catalog No. 503029230
Encompass_Preferred
For Speciality Tube Racks  (Micronic, Fluidx) H: 4″-4 1/2″ (101.5-107.5mm) Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Dimensions H x D x W: 26 5/8 x 3 5/8 x 5 1/2inch (676 x 92 x 140mm)Compartment Dimensions h x d x w: 4 5/16 x 3 7/16 x 5 3/16inch (110 x 88 x 132mm)
Catalog No. 50-302-9230 Supplier Alkali Scientific Supplier No. CFMR6
For Speciality Tube Racks  (Micronic, Fluidx) H: 4″-4 1/2″ (101.5-107.5mm) Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Dimensions H x D x W: 26 5/8 x 3 5/8 x 5 1/2inch (676 x 92 x 140mm)Compartment Dimensions h x d x w: 4 5/16 x 3 7/16 x 5 3/16inch (110 x 88 x 132mm)