Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Rack for SBS Formatted Boxes, Holds 10 Boxes
SDP
Catalog No. 503029337
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 15/16″- 1 1/8″ (23.8-28.45mm) Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Dimensions L x H x W: 12 3/8 x 3 5/8 x 5 1/2 inch ( 315 x 92 x 140mm)Compartment Size l x h x W: 1 3/16 x 3 7/16 x 5 3/16 inch (30 x 88 x 132mm)
Catalog No. 50-302-9337 Supplier Alkali Scientific Supplier No. CFMM10
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 15/16″- 1 1/8″ (23.8-28.45mm) Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Dimensions L x H x W: 12 3/8 x 3 5/8 x 5 1/2 inch ( 315 x 92 x 140mm)Compartment Size l x h x W: 1 3/16 x 3 7/16 x 5 3/16 inch (30 x 88 x 132mm)