Learn More
Alkali Scientific Alkali Scientific™ Vertical Freezer Racks for 100-Cell Hinged Plastic Boxes, Rack Only, 13 Boxes High
SDP
Catalog No. 503028232
Encompass_Preferred
For 100 Cell Hinge Lid Storage Boxes Vertical freezer racks for storage of 100-cell hinged plastic boxes, features a 13 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and features locking rods for added security. Dimensions H x D x W: 30 1/4 x 5 7/8 x 6 1/4 inches (768 x 149 x 159mm)Compartment Dimensions h x d x w: 2 5/16 x 5 9/16 x 6 1/8 Inches ( 58 x 141 x 155mm) *Lock device is ordered separately.
Catalog No. 50-302-8232 Supplier Alkali Scientific Supplier No. VFR13
For 100 Cell Hinge Lid Storage Boxes Vertical freezer racks for storage of 100-cell hinged plastic boxes, features a 13 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and features locking rods for added security. Dimensions H x D x W: 30 1/4 x 5 7/8 x 6 1/4 inches (768 x 149 x 159mm)Compartment Dimensions h x d x w: 2 5/16 x 5 9/16 x 6 1/8 Inches ( 58 x 141 x 155mm) *Lock device is ordered separately.