Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Vertical Freezer Half Rack, Holds (3) 2" Standard Boxes, Rack Dims 16 1/2H x 2 1/4D x 5 9/16W" ( 419 x 57 x 141mm) Compartment Dims 5 3/8H x 2 1/8D x 5 1/4W" (137 x 54.4 x 133mm), With Locking Rod
Catalog No. NC2208078
Encompass_Preferred
Rack Designed to hold (3) Standard 2"Cardboard Boxes, Plastic (Polypropylene or Polycarbonate) or Metal (Stainless Steel or Aluminum) Vertically, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208078 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. LR32
Rack Designed to hold (3) Standard 2"Cardboard Boxes, Plastic (Polypropylene or Polycarbonate) or Metal (Stainless Steel or Aluminum) Vertically, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.