Learn More
OHAUS Vertical Hi-Speed 15 mL Tube Hold
Catalog No. 01804342 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-342 Vortex Attachment
1 options
Catalog No. 01-804-342 Supplier OHAUS Supplier No. 83041244
Add to Cart
Add to Cart
Tube Holder holds six 15 mL tubes vertically.
  • Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse.
  • Optimized for high speed vortexing a small number of tubes.
  • No additional attachment necessary.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse