Learn More
Chemglass Life Sciences Vertical Holder; Holds (6) 50mL, (6) 15mL, and (12) Microtubes (Not Shown)
SDP
Catalog No. 50143314
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-143-314 Supplier Chemglass Life Sciences Supplier No. CLS4891033
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.