Learn More
Sutter Instruments Vertical Micropipette Puller with Platinum/Iridium Filament
SDP
Catalog No. NC9850114
Encompass_Preferred
The model P-30 vertical micropipette puller is designed for the fabrication of basic micropipettes and patch-type pipettes. It will pull micropipettes with tip diameters as small as 0.3μm and moderate taper lengths (6mm to 10mm). By using an includedpatching attachment, the P-30 will pull a patch-type pipette. The platinum filament is more efficient at heating and cooling, and, although it is more prone to damage than the Nichrome filament, the platinum filament is recommended for thick wall and aluminosilicate glass and applications requiring shorter taper lengths.
Catalog No. NC9850114 Supplier Sutter Instruments Supplier No. FGP30P
The model P-30 vertical micropipette puller is designed for the fabrication of basic micropipettes and patch-type pipettes. It will pull micropipettes with tip diameters as small as 0.3μm and moderate taper lengths (6mm to 10mm). By using an includedpatching attachment, the P-30 will pull a patch-type pipette. The platinum filament is more efficient at heating and cooling, and, although it is more prone to damage than the Nichrome filament, the platinum filament is recommended for thick wall and aluminosilicate glass and applications requiring shorter taper lengths.