Learn More
OHAUS Vertical Microtube Hold 24
Catalog No. 01804409 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-409 Vortex Attachment
1 options
Catalog No. 01-804-409 Supplier OHAUS Supplier No. 83041240
Add to Cart
Add to Cart
Tube Holder holds twenty-four microtubes vertically.
  • Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse.
  • No additional attachment necessary.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse