Learn More
Research Products International Corp Vertical High Speed Microtube Holder for 15mL Tubes, 5 Tube Capacity
SDP
Catalog No. 501977836
Encompass
Tube Holder holds six 15mL tubes vertically. Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse. Optimized for high speed vortexing a small number of tubes. No additional attachment necessary.
Catalog No. 50-197-7836 Supplier Research Products International Corp Supplier No. 155328
Tube Holder holds six 15mL tubes vertically. Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse. Optimized for high speed vortexing a small number of tubes. No additional attachment necessary.