Learn More
Chemglass Life Sciences Vertical Microtube Tube Holder (24 Tubes) (Not Shown)
SDP
Catalog No. 5000083173
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-000-83173 Supplier Chemglass Life Sciences Supplier No. CLS4891031
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.