Learn More
Labrepco Inc VERTICAL MINI RACKS FOR CHEST
SDP
Catalog No. 509020156
Encompass_Preferred
Rack, Mini Vertical; Labrepco; Crystal; For Chest Freezers and Liquid Nitrogen Tanks; For 2 in. mini boxes; Locking rod is included; Capacity (Box): 13; Dimensions: 717 x 86 x 82mm
Catalog No. 50-902-0156 Supplier Labrepco Inc Supplier No. MCF132
Rack, Mini Vertical; Labrepco; Crystal; For Chest Freezers and Liquid Nitrogen Tanks; For 2 in. mini boxes; Locking rod is included; Capacity (Box): 13; Dimensions: 717 x 86 x 82mm