Learn More
MSC TPI Vertical Mount Bracket, 25-50 kW, 5100 Series, 1 ea.
Catalog No. 5000508456
Encompass
This vertical mount bracket is designed to support 5100 series fan-forced electric heaters, providing secure placement for overhead installation. It assists in positioning the heating unit away from walls or ceilings to enhance air circulation and operational safety.
  • Compatible with 5100 series fan-forced electric heaters
  • Constructed from heavy 18 gauge steel
  • Finished with powder coated paint for durability
  • Designed for 25 to 50 kW heater units
  • Includes one mounting bracket
Catalog No. 50-005-08456 Supplier MSC Supplier No. 62083340
This vertical mount bracket is designed to support 5100 series fan-forced electric heaters, providing secure placement for overhead installation. It assists in positioning the heating unit away from walls or ceilings to enhance air circulation and operational safety.
  • Compatible with 5100 series fan-forced electric heaters
  • Constructed from heavy 18 gauge steel
  • Finished with powder coated paint for durability
  • Designed for 25 to 50 kW heater units
  • Includes one mounting bracket