Learn More
MSC TPI Corporation Vertical Mount Bracket, 60 to 100 kW, 5100 Series, 1 ea
Catalog No. 5000506978
Encompass
This vertical mount bracket is designed to support 5100 series unit heaters with power ratings between 60 kW and 100 kW. It facilitates vertical discharge installations for suspended, fan-forced unit heating systems.
  • Compatible with 5100 series unit heaters ranging from 60 kW to 100 kW
  • Engineered for vertical discharge mounting configurations
  • Durable construction suitable for industrial heater support
  • Designed for use with suspended fan-forced units
Catalog No. 50-005-06978 Supplier MSC Supplier No. 72353543
This vertical mount bracket is designed to support 5100 series unit heaters with power ratings between 60 kW and 100 kW. It facilitates vertical discharge installations for suspended, fan-forced unit heating systems.
  • Compatible with 5100 series unit heaters ranging from 60 kW to 100 kW
  • Engineered for vertical discharge mounting configurations
  • Durable construction suitable for industrial heater support
  • Designed for use with suspended fan-forced units