Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Vertical Mounting Blocks, 2" 6 Position
SDP
Supplier:  Electron Microscopy Sciences 5015402
Encompass
The blocks are used with the Model 155D Lapping and Polishing fixture for edge polishing 2" (50mm) diameter mounted blocks. It accommodates vertically mounted samples. Star Pattern, Reversible, 6-position (Max 20mm Specimen Length), Stainless Steel.
Catalog No. 50-980-987