Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Vertical Mounting Blocks, 2" 6 Position
SDP
Catalog No. 50980988
Encompass
The blocks are used with the Model 155D Lapping and Polishing fixture for edge polishing 2" (50mm) diameter mounted blocks. It accommodates vertically mounted samples. 6-position for 12mm Diameter Rod or 10mm Cube (Max 20mm Specimen Length), Stainless Steel.
Catalog No. 50-980-988 Supplier Electron Microscopy Sciences Supplier No. 5015403
The blocks are used with the Model 155D Lapping and Polishing fixture for edge polishing 2" (50mm) diameter mounted blocks. It accommodates vertically mounted samples. 6-position for 12mm Diameter Rod or 10mm Cube (Max 20mm Specimen Length), Stainless Steel.