Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Vertical Mounting Blocks, 2" 4 Position
SDP
Catalog No. 50980986
Encompass
The blocks are used with the Model 155D Lapping and Polishing fixture for edge polishing 2" (50mm) diameter mounted blocks. It accommodates vertically mounted samples. Star Pattern, 4-Position. Stainless Steel.
Catalog No. 50-980-986 Supplier Electron Microscopy Sciences Supplier No. 5015401
The blocks are used with the Model 155D Lapping and Polishing fixture for edge polishing 2" (50mm) diameter mounted blocks. It accommodates vertically mounted samples. Star Pattern, 4-Position. Stainless Steel.