Learn More
World Precision Instrument Vertical Puller-SU-P30N
SDP
Catalog No. 502538515
Encompass
The model SU-P30 vertical micropipette puller is designed for the fabrication of basic micropipettes and patch-type pipettes. It will pull micropipettes with tip diameters as small as 0.3 µm and moderate taper lengths (6 to 10 mm). By using an included patching attachment, the puller will pull the standard double pull patch-type pipette. Used with thin wall glass capillaries, it will generate pipettes or needles suitable for microinjection studies. The puller is ideal for student laboratories and other situations which call for an economical, reliable pipette pulling device.
View our Micropipette Puller Comparison Chart
Catalog No. 50-253-8515 Supplier World Precision Instrument Supplier No. SUP30N
Add to Cart
Add to Cart
Request a Quote
The model SU-P30 vertical micropipette puller is designed for the fabrication of basic micropipettes and patch-type pipettes. It will pull micropipettes with tip diameters as small as 0.3 µm and moderate taper lengths (6 to 10 mm). By using an included patching attachment, the puller will pull the standard double pull patch-type pipette. Used with thin wall glass capillaries, it will generate pipettes or needles suitable for microinjection studies. The puller is ideal for student laboratories and other situations which call for an economical, reliable pipette pulling device.
View our Micropipette Puller Comparison Chart