Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Vertical Freezer Rack Holds (9) 2" Mini Boxes, Rack Dims 20 1/16 H x 3 3/8W x 3 1/4D" (510 x 86 x 82mm) with Compartment Dims 2 13/16H x 3 1/16W x 3 1/16D" (55 x 78 x 78mm), With Locking Rod
Catalog No. NC2208035
Encompass_Preferred
Rack Designed to hold (9) Standard 2"Mini Cardboard Boxes and Polycarbonate Boxes, for use in Chest Freezers and Liquid Nitrogen Tanks, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Designs are available. Security Lock Devices are Available for each Rack.
This item is not returnable. View return policy
Catalog No. NC2208035 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. MCF92
Rack Designed to hold (9) Standard 2"Mini Cardboard Boxes and Polycarbonate Boxes, for use in Chest Freezers and Liquid Nitrogen Tanks, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Designs are available. Security Lock Devices are Available for each Rack.