Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Vertical Freezer Rack Holds (5) 3.75" Standard Boxes, Rack Dims 20 1/4H x 5 5/8W x 5 1/2W" (514 x 143 x 140mm) with Compartment Dims 3 15/16H x 5 1/4W x 5 3/8D" (100 x 134 x 136mm), with Locking Rod
Catalog No. NC2208036
Encompass_Preferred
Rack Designed to hold (5) 3.75"H Cardboard Boxes or Polycarbonate Boxes for use in Chest Freezers and Liquid Nitrogen Tanks, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208036 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. CF5375
Rack Designed to hold (5) 3.75"H Cardboard Boxes or Polycarbonate Boxes for use in Chest Freezers and Liquid Nitrogen Tanks, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.