Learn More
Alkali Scientific Vertical Rack for 3.75 boxes h
SDP
Supplier:  Alkali Scientific CF6375
Encompass_Preferred
The Vertical Rack for 3 75-inch High Boxes is a specially designed storage solution tailored for accommodating larger storage boxes within laboratory freezers including chest and upright models as well as liquid nitrogen tanks This rack is crafted to meet the specific needs of laboratories that require additional height clearance for their unique storage boxes ensuring both optimal space utilization and easy access to stored samples Product Features Durable Construction Made from materials selected for their strength and durability this rack is built to withstand the extreme temperatures and conditions of laboratory freezers and liquid nitrogen tanks Its construction ensures long-term reliability and stability for your stored samples Efficient Space Utilization By taking advantage of vertical space this rack maximizes storage capacity within your freezer or tank It allows for the organized stacking of large boxes making efficient use of available space and facilitating better inve
Catalog No. 50-302-6408