Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for 50-Cell 0.5 ml Microtube Boxes, Holds 16 boxes
SDP
Catalog No. 503029355
Encompass_Preferred
For 50-Cell 0.5 ml Storage Boxes Dimensions H x D x W: 18 1/8 x 5 1/2 x 5 1/2inch (460 x 140 x 140 mm)Compartment Dimensions h x d x w: 2 3/16 x 5 5/16 x 5 1/8 inch (56.3 x 135 x 130mm) *Security lock device is ordered separately.
Catalog No. 50-302-9355 Supplier Alkali Scientific Supplier No. CFM805
For 50-Cell 0.5 ml Storage Boxes Dimensions H x D x W: 18 1/8 x 5 1/2 x 5 1/2inch (460 x 140 x 140 mm)Compartment Dimensions h x d x w: 2 3/16 x 5 5/16 x 5 1/8 inch (56.3 x 135 x 130mm) *Security lock device is ordered separately.