Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for 50-Cell 1.5 ml Microtube Boxes, Holds 12 boxes
SDP
Catalog No. 503027232
Encompass_Preferred
For 50-Cell 1.5 ml Storage Boxes Dimensions H x D x W: 14 1/16 x 5 7/8 x 6 1/4 inch (357 x 149 x 159mm)|Compartment Size h x d x w: 2 5/16 x 5 1/2 x 5 15/16 (58 x 140 x 150.7mm) *Security lock device is ordered separately.
Catalog No. 50-302-7232 Supplier Alkali Scientific Supplier No. CFM615
For 50-Cell 1.5 ml Storage Boxes Dimensions H x D x W: 14 1/16 x 5 7/8 x 6 1/4 inch (357 x 149 x 159mm)|Compartment Size h x d x w: 2 5/16 x 5 1/2 x 5 15/16 (58 x 140 x 150.7mm) *Security lock device is ordered separately.