Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for 96-Well Microtube Boxes, Holds 10 Boxes
SDP
Catalog No. 503029367
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″- 2 1/82″ (17.3-53.5mm) The CFTB-10 vertical freezer racks for 96-Well Microtube Boxes, features a 10 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and also features locking rods for added security.  Dimensions (DxWxH): 3 5/8 x 5 1/2 x 22 3/8 inches (92 x 140 x 568mm)Compartment Dimensions: 2 3/16 x 3 7/16 x 5 3/16 inch ( 55.6 x 88 x 131.5mm) *Security lock device is ordered separately.
Catalog No. 50-302-9367 Supplier Alkali Scientific Supplier No. CFTB10
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″- 2 1/82″ (17.3-53.5mm) The CFTB-10 vertical freezer racks for 96-Well Microtube Boxes, features a 10 boxes high configuration for storage in a laboratory chest or LN2 freezer. These vertical freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer and also features locking rods for added security.  Dimensions (DxWxH): 3 5/8 x 5 1/2 x 22 3/8 inches (92 x 140 x 568mm)Compartment Dimensions: 2 3/16 x 3 7/16 x 5 3/16 inch ( 55.6 x 88 x 131.5mm) *Security lock device is ordered separately.