Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for SBS formatted boxes, Holds 4 boxes
SDP
Catalog No. 503029378
Encompass_Preferred
For Speciality Tube Racks  (Micronic, Fluidx) H: 3 7/16″ (88.5mm) Dimensions H x D x W: 14 3/4 x 3 5/8 x 5 1/2inch (374 x 92 x 140mm)Compartment Dimensions h x d x w: 3 9/16 x 3 7/16 x 5 3/16inch (91 x 88 x 132mm)
Catalog No. 50-302-9378 Supplier Alkali Scientific Supplier No. CFMH4
For Speciality Tube Racks  (Micronic, Fluidx) H: 3 7/16″ (88.5mm) Dimensions H x D x W: 14 3/4 x 3 5/8 x 5 1/2inch (374 x 92 x 140mm)Compartment Dimensions h x d x w: 3 9/16 x 3 7/16 x 5 3/16inch (91 x 88 x 132mm)