Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for SBS formatted boxes, Holds 6 boxes
SDP
Catalog No. 503029412
Encompass_Preferred
For Speciality Tube Racks  (Micronic, Fluidx) H: 3 7/16″ (88.5mm) Dimensions H x D x W: 21 15/16 x 3 5/8 x 5 1/2inch (557 x 92 x 140mm)Compartment Dimensions h x d x w: 3 9/16 x 3 7/16 x 5 3/16inch (91 x 88 x 132mm)
Catalog No. 50-302-9412 Supplier Alkali Scientific Supplier No. CFMH6
For Speciality Tube Racks  (Micronic, Fluidx) H: 3 7/16″ (88.5mm) Dimensions H x D x W: 21 15/16 x 3 5/8 x 5 1/2inch (557 x 92 x 140mm)Compartment Dimensions h x d x w: 3 9/16 x 3 7/16 x 5 3/16inch (91 x 88 x 132mm)