Learn More
Alkali Scientific Vertical Racks for 96-Deep-Wel
SDP
Supplier:  Alkali Scientific CFDP11
Encompass_Preferred
For 96-Deep Well Microtiter PlatesFor Speciality Tube Racks (Matrix Micronic Fluidx Abgene) H 9/16- 1 3/4 (14 44-44 32mm) High quality vertical racks for chest freezers or liquid nitrogen tanks are made of corrosion resistant stainless steel Security lock devices are also available for each rack Use the internal dimensions obtained for your chest freezer or liquid nitrogen tank (fig 2 or fig 3 on this page) to determine the maximum height (H) and number of racks you need for your chest freezer or liquid nitrogen tank Vertical racks are sold with or without standard 2in cardboard polycarbonate or metal boxes All racks include stainless steel locking rods Our Freezer Racks have a variety applications for scientific research clinical pharmaceutical and other fields This series of products are used for optimizing space reliable sample storage in freezer and liquid nitrogen tanks They fit all models of Freezers liquid nitrogen tanks and blood bag storage systems The innovative d
Catalog No. 50-302-9196