Learn More
Labrepco Inc VERTICAL RACKS FOR CHEST FREEZ
SDP
Supplier:  Labrepco Inc CF102
Encompass_Preferred
Rack, Vertical; Labrepco; Crystal; For Chest Freezer and Liquid Nitrogen Tanks; For standard 2 in. boxes; Includes locking rod; Capacity (Box): 10; Dimensions: 559 x 143 x 140mm
Catalog No. 50-902-0131