Learn More
Labrepco Inc VERTICAL RACKS FOR CHEST FREEZ
SDP
Catalog No. 509020133
Encompass_Preferred
Rack, Vertical; Labrepco; Crystal; For Chest Freezer and Liquid Nitrogen Tanks; For standard 2 in. boxes; Includes locking rod; Capacity (Box): 12; Dimensions: 670 x 143 x 140mm
Catalog No. 50-902-0133 Supplier Labrepco Inc Supplier No. CF122
Rack, Vertical; Labrepco; Crystal; For Chest Freezer and Liquid Nitrogen Tanks; For standard 2 in. boxes; Includes locking rod; Capacity (Box): 12; Dimensions: 670 x 143 x 140mm