Learn More
Labrepco Inc VERTICAL RACKS FOR CHEST FREEZ
Supplier: Labrepco Inc CF93
Rack, Vertical; Labrepco; Crystal; For Chest Freezer and Liquid Nitrogen Tanks; For standard 3 in. boxes; Includes locking rod; Capacity (Box): 9; Dimensions: 717 x 143 x 140mm