Learn More
Grainger SARGENT Surface Vertical Rod Exit Device, Bronze, 36 in, Classroom, 1 ea
Catalog No. 502787237
Encompass_Preferred
This heavy-duty, Grade 1 surface vertical rod exit device is engineered for demanding environments requiring reliable security and durability. Designed for use on 1-3/4 in. thick wood or metal doors, this classroom-function device features two-point latching that is conveniently adjustable through the center case. The pushpad touchbar offers smooth operation for quick exit, while the unit is built to withstand high-traffic, abusive applications over the life of the building.
  • ANSI/BHMA Grade 1 certified for strength and cycle durability
  • Designed for wide stile applications with 4-1/2 in. minimum stile width
  • Classroom function with hex key dogging feature
  • Right hand reverse orientation
  • Dark oxidized satin bronze, oil-rubbed architectural finish
  • Includes wood and machine screws for standard installation
  • UL 305 panic listed for safety
Catalog No. 50-278-7237 Supplier Grainger Supplier No. 18F608
This heavy-duty, Grade 1 surface vertical rod exit device is engineered for demanding environments requiring reliable security and durability. Designed for use on 1-3/4 in. thick wood or metal doors, this classroom-function device features two-point latching that is conveniently adjustable through the center case. The pushpad touchbar offers smooth operation for quick exit, while the unit is built to withstand high-traffic, abusive applications over the life of the building.
  • ANSI/BHMA Grade 1 certified for strength and cycle durability
  • Designed for wide stile applications with 4-1/2 in. minimum stile width
  • Classroom function with hex key dogging feature
  • Right hand reverse orientation
  • Dark oxidized satin bronze, oil-rubbed architectural finish
  • Includes wood and machine screws for standard installation
  • UL 305 panic listed for safety