This surface-mounted vertical rod exit device provides secure egress and is designed for use on doors up to 36 in wide. Featuring stainless steel construction, this heavy-duty hardware is suitable for demanding commercial environments and meets rigorous safety and performance standards, including fire rating requirements for up to 3-hour fire doors.
- ANSI/BHMA Grade 1 certified for operational and security performance
- UL and cUL listed for panic and fire exit hardware (UL10C)
- Surface-mounted vertical rod design connects to top and bottom door latches
- Features hex key dogging for versatile operation
- Meets ADA accessibility standards and NFPA/IBC egress requirements
- Durable stainless steel construction with satin finish
- Compatible with standard commercial door frames
- Meets windstorm resistant requirements including Florida Building Code FL2998