Learn More
Grainger Sargent Surface Vertical Rod Exit Device, Brass, Grade 1, 1 ea
Catalog No. 502786362
Encompass_Preferred
The 8700 Series Surface Vertical Rod Exit Device is a robust, Grade 1 mechanical exit solution designed for security and fire safety. It features two-point latching at the top and bottom of the door, with adjustments conveniently accessible through the center case. This unit is constructed from brass and engineered for reliable operation in demanding environments.
  • Features two-point latching mechanism for enhanced security
  • Adjustable through the center case for simplified installation
  • Fire-rated per UL10C standards
  • ADA compliant design
  • Vandal resistant construction
  • Includes hex key dogging for lockdown capability
  • Suitable for doors with 1 3/4 in. thickness
Catalog No. 50-278-6362 Supplier Grainger Supplier No. 18F594
The 8700 Series Surface Vertical Rod Exit Device is a robust, Grade 1 mechanical exit solution designed for security and fire safety. It features two-point latching at the top and bottom of the door, with adjustments conveniently accessible through the center case. This unit is constructed from brass and engineered for reliable operation in demanding environments.
  • Features two-point latching mechanism for enhanced security
  • Adjustable through the center case for simplified installation
  • Fire-rated per UL10C standards
  • ADA compliant design
  • Vandal resistant construction
  • Includes hex key dogging for lockdown capability
  • Suitable for doors with 1 3/4 in. thickness