Learn More
Electron Microscopy Sciences Secador™ Vertical Shelves for Secador Desiccator Cabinets
SDP
Catalog No. 501899535
Encompass_Preferred
To convert your Model 4.0 Secador Autodesiccator Cabinets or Secador Desiccator Cabinets from the Horizontal configuration to the Vertical, or from Vertical configuration to Horizontal, all you need is a new set of shelves. 3 shelves.
Catalog No. 50-189-9535 Supplier Electron Microscopy Sciences Supplier No. 7122920
To convert your Model 4.0 Secador Autodesiccator Cabinets or Secador Desiccator Cabinets from the Horizontal configuration to the Vertical, or from Vertical configuration to Horizontal, all you need is a new set of shelves. 3 shelves.