Learn More
Electron Microscopy Sciences Vertical Staining Jar 4 per pack
SDP
Catalog No. 501899241
Encompass_Preferred
An all glass rectangular staining jar which accommodates 8 to 16 slides and comes with a glass lid. Its wide top eliminates the need for forceps in the handling of the slides. It accommodates slides up to 4"x1 1/2" (103 mm x 39 mm) in size.
Catalog No. 50-189-9241 Supplier Electron Microscopy Sciences Supplier No. 7031804
An all glass rectangular staining jar which accommodates 8 to 16 slides and comes with a glass lid. Its wide top eliminates the need for forceps in the handling of the slides. It accommodates slides up to 4"x1 1/2" (103 mm x 39 mm) in size.