Learn More
Newcomer Supply Staining Rack for 30 Slide Staining Dish
SDP
Catalog No. NC0779540
Encompass
The Stainless Steel Staining Rack accommodates 30 vertical slides. Outside Dimensions: 2 7/8" x 2 3/4" x 3" (H). Racks are also available in 2 other sizes to accommodate 20 and 60 slides.
Catalog No. NC0779540 Supplier Newcomer Supply Supplier No. 6817B
The Stainless Steel Staining Rack accommodates 30 vertical slides. Outside Dimensions: 2 7/8" x 2 3/4" x 3" (H). Racks are also available in 2 other sizes to accommodate 20 and 60 slides.