Learn More
Newcomer Supply Staining Rack for 60 Slide Staining Dish
SDP
Catalog No. NC1575383
Encompass
The Stainless Steel Staining Rack accommodates 60 vertical slides. Outside Dimensions: 5 1/4" x 3 1/8" x 3" (H). Racks are also available in 2 other sizes to accommodate 20 and 30 slides.
Catalog No. NC1575383 Supplier Newcomer Supply Supplier No. 6817C
The Stainless Steel Staining Rack accommodates 60 vertical slides. Outside Dimensions: 5 1/4" x 3 1/8" x 3" (H). Racks are also available in 2 other sizes to accommodate 20 and 30 slides.