Learn More
Andwin Scientific VERTICAL STORAGE BINS. COMPART
SDP
Catalog No. 50820164
Encompass_Preferred
The vertical storage bin can accommodate approximately 200 microcentrifuge (1.5 ml) tubes. There are three compartments that can be easily accessed with one hand and filled through the top hinged lid. This product provides a dust-free storage. This measures 11 1/2" W x 6 1/2" D x 8 1/2" H.
Catalog No. 50-820-164 Supplier Andwin Scientific Supplier No. 30820300
The vertical storage bin can accommodate approximately 200 microcentrifuge (1.5 ml) tubes. There are three compartments that can be easily accessed with one hand and filled through the top hinged lid. This product provides a dust-free storage. This measures 11 1/2" W x 6 1/2" D x 8 1/2" H.