Learn More
Analytical Sales & Services Vertical Storage Rack for Up To Eight 80-well Microtube Racks
Catalog No. 5000805795
Encompass
Storage Rack for Up To Eight 80-well Microtube Racks Vertical Rack, For Ambient Termperature Only - Do Not Refrigerate
Catalog No. 50-008-05795 Supplier Analytical Sales & Services Supplier No. STRV80
Storage Rack for Up To Eight 80-well Microtube Racks Vertical Rack, For Ambient Termperature Only - Do Not Refrigerate