Learn More
Electron Microscopy Sciences SEM Sample Holder Vertical for Thin Sample With Pin A, 25 mm dia and 10 mm thick
SDP
Catalog No. 5028716
Encompass_Preferred
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. Holds thin samples vertically in the SEM or microscope. Useful for cross sections of silicon wafers or multilayer capicators. Pin overall measures: 28mm L x3.1mm dia
Catalog No. 50-287-16 Supplier Electron Microscopy Sciences Supplier No. 7591501
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. Holds thin samples vertically in the SEM or microscope. Useful for cross sections of silicon wafers or multilayer capicators. Pin overall measures: 28mm L x3.1mm dia