Learn More
Labrepco Inc VERTICLE RACKS 2INBOX/14 BOXES
SDP
Catalog No. NC1685998
Encompass_Preferred
Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel. Racks are made for standard 2in boxes, 14 boxes. All racks include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.*Rack only; Boxes and cell dividers not included.*Lock Device Option: DR-31 (ordered separately)
Catalog No. NC1685998 Supplier Labrepco Inc Supplier No. CF142
Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel. Racks are made for standard 2in boxes, 14 boxes. All racks include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.*Rack only; Boxes and cell dividers not included.*Lock Device Option: DR-31 (ordered separately)